27 May 2026
LALI-ToFSIMS
ToFSIMS collection of visualization modules for Exum Massbox mass spectrometer HDF5 data,
LALISnap
Time-of-flight secondary ion mass spectrometry (ToF-SIMS) employs a trusted spectroscopic technique which collects and separates isotopes according to their mass-to-charge ratio (m/z).
ExumMassBoxViz module reads back specific element(s) defined by atomic mass from a set of HDF5 files produced by Exum Massbox mass spectrometer..
ToFSIMSFindExumPeaks  module finds most prominent elements in the Peak Data table