16 Oct 2019
PerGeos TCL script for processing all files in a folder with a recipe
Process all files in a folder by applying the same recipe to all of them individually.
This script applies the same recipe to all the data listed in a
file folder, and save the recipe result for each of them in a folder called "results".
It can be used as a basis of an automation routine. The recipe can
be changed with the port "recipeFileName", and the input file folder can be
changed with the port "Data Directory".
The results are saved as Avizo binary files, assuming that the
recipe produces a binary output.
Note : multiple outputs are possible, including snapshots,
spreadsheets, but are not exposed in the script itself.