3 Jul 2020
Porosity and Permeability by blocks
Splits a binary pore space into blocks and output one porosity and permeability value for every block
This is an Xtra TCL script that splits a binary pore space into blocks and output one porosity and permeability value for every block.
Choose the appropriate binary pore
space in the data port, and indicate how many blocks you want in X, Y and Z.
Check
if you need both porosity and permeability to be computed for each block, and
press Apply.
After the computation is done, you get 3 outputs :
- a spreadsheet with the block descriptions and the computed values
- the porosity by block data
- the permeability by block data