16 Oct 2019
Refine Surface
This module refines a surface.
refinesurface2022
This module refines a surface by replacing qualified triangles with 4 smaller triangles. A triangle gets qualified for refinement if it's shortest edge is superior to a user defined minimum length, and if it's longest edge is superior to a user defined maximum length.
The algorithm iterates through all triangles until no more triangles are qualified for refinement, or until a maximum user defined number of iterations is reached.
Note: Unlike other approaches, this algorithm does not smooth edges because the replaced triangles inherit their orientation from their parent triangle.
Download a version compatible with Amira-Avizo 2022.1 HERE, 2022.2 HERE, 2023.1 HERE, 2023.2 HERE.
To obtain a version of this Xtra compatible with a more recent version of Amira-Avizo, please contact us via our Customer Support Center.