13 May 2022
Scientific Scalebar
This Xtra automatically sets the scalebar parameters to comply with the requirements for scientific papers.
5-13-2022 3-02-28 PM
The Xtra wraps a Scalebars module and sets its parameters to be the most compliant possible with the requirements for scientific papers. It also exposes only the relevant parameters in order to ease their tuning and make the user interface of the module easier to use.
You can create the module by right-clicking on your data, then go to Xtra>Annotations.