18 Oct 2022
Surface Material Grid View
This Xtra arranges all materials of a surface in a grid view.
The script arranges all materials of a surface in a grid view (displayed in [xy] plane).
The spacing, the origin and the number of columns in x direction can be specified.
If a Label Analysis has been computed for the materials, materials can be sorted using the Label Analysis measures as criteria.
This Xtra is very useful, e.g. when visually inspecting the different materials inside a volume.
To create the Xtra, right-click on a surface and search for Xtra > Surface Material Grid View.