Poloxamer 188 Non-ionic Surfactant (10%) - FAQs
View additional product information for Poloxamer 188 Non-ionic Surfactant (10%) - FAQs (24040032)
1 product FAQs found
What is Pluronic F-68 Non-ionic Surfactant (100X) dissolved in?
It is a 10% solution of Pluronic F-68 in distilled water.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.