NanoDrop™ CF-8 Calibration Check Kit - FAQs
View additional product information for NanoDrop™ Reagents - FAQs (CHEM-PV-8, CHEM-CF-8, CHEM-PR1-KIT, CHEM-PV-1, CHEM-CF-1)
2 product FAQs found
How often should I check the calibration of my NanoDrop 8000 Spectrophotometer?
We recommend you check the calibration of your NanoDrop 8000 Spectrophotometer every 6 months using the NanoDrop CF-8 Calibration Check Kit.
How do I check the calibration of my NanoDrop 8000 Spectrophotometer? Do I need to calibrate it?
You can easily check the calibration of your NanoDrop 8000 Spectrophotometer by using the NanoDrop CF-8 Calibration Check Kit, which contains two CF-1 Ampules and two 8-well PCR tube strips. The CF-8 kit is available from the Thermo Fisher website (Cat. No. CHEM-CF-8).
If re-calibration is ever needed, it is performed by a trained service technician. Due to the precise nature of the NanoDrop instrument design, these spectrophotometers rarely require re-calibration.