Search
View additional product information for NCode™ miRNA First-Strand cDNA Synthesis Kit - FAQs (MIRC50)
1 product FAQs found
You can store your cDNA at 2-6 degrees C for up to 24 hours. For long-term storage, store the cDNA at -15 to -25 degrees C and add EDTA to a final concentration of 1 mM to prevent degradation.