Search
Search

CESA2| Gene Symbol: CESA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120095.4 | NP_195645.1 | 7 - 8 | 2200 | 65 | |
| GenBank mRNA | AF027173.2 | - | 7 - 8 | 1680 | 65 | |
| KJ139038.1 | - | 7 - 8 | 1510 | 65 | ||
Set Membership: |
|