Search
Search

ESP4| Gene Symbol: ESP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342587.1 | NP_001330013.1 | 23 - 24 | 3765 | 72 | |
| NM_001342588.1 | NP_001330014.1 | 24 - 25 | 3722 | 72 | ||
| NM_120218.2 | NP_195760.1 | 24 - 25 | 3507 | 72 | ||
Set Membership: |
|