Search
Search

PBB2| Gene Symbol: PBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203514.2 | NP_001190443.1 | 5 - 6 | 541 | 100 | |
| NM_001344344.1 | NP_001318715.1 | 5 - 6 | 667 | 100 | ||
| NM_123422.4 | NP_198874.1 | 5 - 6 | 590 | 100 | ||
| NM_180777.3 | NP_851108.1 | 5 - 6 | 649 | 100 | ||
| GenBank mRNA | AF043531.1 | - | 5 - 6 | 491 | 100 | |
Set Membership: |
|