Search
Search

KAT5| Gene Symbol: KAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036960.2 | NP_001032037.1 | 4 - 5 | 314 | 93 | |
| NM_001344808.1 | NP_001331840.1 | 4 - 5 | 486 | 93 | ||
| NM_124265.5 | NP_568704.2 | 3 - 4 | 1181 | 93 | ||
| NM_180826.4 | NP_851157.1 | 3 - 4 | 331 | 93 | ||
| GenBank mRNA | AF062590.1 | - | 3 - 4 | 211 | 93 | |
| AF062591.1 | - | 3 - 4 | 430 | 93 | ||
| AY079026.1 | - | 3 - 4 | 316 | 93 | ||
| AY136455.1 | - | 3 - 4 | 219 | 93 | ||
| BT002594.1 | - | 2 - 3 | 118 | 93 | ||
Set Membership: |
|