Search
Search

CYL1| Gene Symbol: CYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343290.1 | NP_001329969.1 | 1 - 2 | 273 | 93 | |
| NM_121372.3 | NP_196873.1 | 1 - 2 | 273 | 93 | ||
| GenBank mRNA | AY080811.1 | - | 1 - 2 | 273 | 93 | |
| AY117179.1 | - | 1 - 2 | 219 | 93 | ||
Set Membership: |
|