Search
Search

ARV2| Gene Symbol: ARV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340322.1 | NP_001328722.1 | 1 - 2 | 137 | 104 | |
| NM_001340323.1 | NP_001328723.1 | 1 - 2 | 137 | 104 | ||
| NM_001340324.1 | NP_001328724.1 | 1 - 2 | 137 | 104 | ||
| NM_001340325.1 | NP_001328720.1 | 1 - 2 | 118 | 104 | ||
| NM_001340326.1 | NP_001328725.1 | 1 - 2 | 118 | 104 | ||
| NM_116381.4 | NP_192060.2 | 1 - 2 | 137 | 104 | ||
| GenBank mRNA | AY758071.1 | - | 1 - 2 | 118 | 104 | |
Set Membership: |
|