Search
Search

AGY1| Gene Symbol: AGY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203724.1 | NP_001190653.1 | 5 - 6 | 1065 | 77 | |
| NM_001340335.1 | NP_001328302.1 | 5 - 6 | 1295 | 77 | ||
| NM_116410.2 | NP_192089.1 | 5 - 6 | 987 | 77 | ||
Set Membership: |
|