Search
Search

AT1G56460| Gene Symbol: AT1G56460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198322.1 | NP_001185251.1 | 4 - 5 | 805 | 83 | |
| NM_001333782.1 | NP_001320733.1 | 3 - 4 | 718 | 83 | ||
| NM_104524.4 | NP_176041.2 | 3 - 4 | 702 | 83 | ||
| GenBank mRNA | AK117690.1 | - | 3 - 4 | 620 | 83 | |
Set Membership: |
|