Search
Search

AT1G63420| Gene Symbol: AT1G63420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334092.1 | NP_001320907.1 | 3 - 4 | 1091 | 71 | |
| NM_105021.3 | NP_176531.1 | 3 - 4 | 1091 | 71 | ||
| GenBank mRNA | KJ138955.1 | - | 3 - 4 | 897 | 71 | |
Set Membership: |
|