Search
Search

AT4G15420| Gene Symbol: AT4G15420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341043.1 | NP_001329347.1 | 1 - 2 | 599 | 106 | |
| NM_117631.3 | NP_567465.1 | 1 - 2 | 276 | 106 | ||
| GenBank mRNA | AF446353.1 | - | 1 - 2 | 264 | 106 | |
| AY086035.1 | - | 1 - 2 | 276 | 106 | ||
| AY143927.1 | - | 1 - 2 | 179 | 106 | ||
| DQ778953.1 | - | 1 - 2 | 220 | 106 | ||
Set Membership: |
|