Search
Search

PAO4| Gene Symbol: PAO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105256.4 | NP_176759.1 | 8 - 9 | 1584 | 82 | |
| GenBank mRNA | AF364953.1 | - | 8 - 9 | 1538 | 82 | |
| BT000353.1 | - | 8 - 9 | 1169 | 82 | ||
Set Membership: |
|