Search
Search

AT1G69710| Gene Symbol: AT1G69710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334419.1 | NP_001322526.1 | 5 - 6 | 972 | 81 | |
| NM_105639.4 | NP_177129.2 | 6 - 7 | 759 | 81 | ||
Set Membership: |
|