Search
Search

ABCC13| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335345.1 | NP_001323939.1 | 22 - 23 | 2414 | 97 | |
| NM_001335346.1 | NP_001323940.1 | 22 - 23 | 2414 | 97 | ||
| NM_001335347.1 | NP_001323936.1 | 21 - 22 | 2331 | 97 | ||
| NM_001335348.1 | NP_001323937.1 | 21 - 22 | 2260 | 97 | ||
| NM_001335349.1 | NP_001323938.1 | 19 - 20 | 2191 | 97 | ||
| NM_126770.8 | NP_178811.7 | 21 - 22 | 2271 | 97 | ||
Set Membership: |
|