Search
Search

AT4G26310| Gene Symbol: AT4G26310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203916.1 | NP_001190845.1 | 2 - 3 | 428 | 66 | |
| NM_118764.3 | NP_567743.1 | 2 - 3 | 633 | 66 | ||
| GenBank mRNA | AY054472.1 | - | 2 - 3 | 295 | 66 | |
| AY093290.1 | - | 2 - 3 | 256 | 66 | ||
Set Membership: |
|