Search
Search

AT3G48380| Gene Symbol: AT3G48380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203103.1 | NP_001190032.1 | 6 - 7 | 1207 | 69 | |
| NM_148861.4 | NP_680114.1 | 6 - 7 | 1207 | 69 | ||
| NM_202673.1 | NP_974402.1 | 6 - 7 | 1184 | 69 | ||
Set Membership: |
|