Search
Search

AT4G29440| Gene Symbol: AT4G29440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036671.1 | NP_001031748.1 | 2 - 3 | 344 | 113 | |
| NM_119089.3 | NP_194673.2 | 2 - 3 | 418 | 113 | ||
Set Membership: |
|