Search
Search

GIP1| Gene Symbol: GIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112163.6 | NP_187929.4 | 1 - 2 | 256 | 90 | |
| GenBank mRNA | AY063883.1 | - | 1 - 2 | 215 | 90 | |
| AY142588.1 | - | 1 - 2 | 184 | 90 | ||
Set Membership: |
|