Search
Search

SWEET16| Gene Symbol: SWEET16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338249.1 | NP_001319571.1 | 2 - 3 | 169 | 66 | |
| NM_001338250.1 | NP_001325596.1 | 2 - 3 | 169 | 66 | ||
| NM_112542.5 | NP_188291.3 | 2 - 3 | 192 | 66 | ||
| GenBank mRNA | AK222159.1 | - | 2 - 3 | 116 | 66 | |
| BT010528.1 | - | 2 - 3 | 73 | 66 | ||
Set Membership: |
|