Search
Search

WNK6| Gene Symbol: WNK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035646.2 | NP_001030723.1 | 5 - 6 | 534 | 151 | |
| NM_001202999.2 | NP_001189928.1 | 4 - 5 | 536 | 151 | ||
| NM_001338354.1 | NP_001319584.1 | 4 - 5 | 709 | 151 | ||
| NM_001338355.1 | NP_001326381.1 | 3 - 4 | 781 | 151 | ||
| NM_001338356.1 | NP_001326380.1 | 4 - 5 | 393 | 151 | ||
| NM_112761.3 | NP_188505.1 | 4 - 5 | 470 | 151 | ||
| GenBank mRNA | AB084270.1 | - | 3 - 4 | 351 | 151 | |
| AK175997.1 | - | 4 - 5 | 470 | 151 | ||
| BT030341.1 | - | 3 - 4 | 351 | 151 | ||
Set Membership: |
|