Search
Search

CST| Gene Symbol: CST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203999.1 | NP_001190928.1 | 3 - 4 | 801 | 86 | |
| NM_119725.5 | NP_195285.3 | 3 - 4 | 847 | 86 | ||
| GenBank mRNA | AY065403.1 | - | 3 - 4 | 636 | 86 | |
| AY096501.1 | - | 3 - 4 | 552 | 86 | ||
Set Membership: |
|