Search
Search

RLP1| Gene Symbol: RLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160845.2 | NP_001154317.1 | 5 - 6 | 636 | 138 | |
| NM_001160846.1 | NP_001154318.1 | 5 - 6 | 684 | 138 | ||
| NM_001331697.1 | NP_001322741.1 | 5 - 6 | 955 | 138 | ||
| NM_001331698.1 | NP_001322740.1 | 5 - 6 | 955 | 138 | ||
| NM_100613.5 | NP_172219.5 | 5 - 6 | 684 | 138 | ||
Set Membership: |
|