Search
Search

PMH2| Gene Symbol: PMH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113131.5 | NP_188872.2 | 4 - 5 | 1163 | 142 | |
| GenBank mRNA | AY062502.1 | - | 4 - 5 | 1055 | 142 | |
| AY093256.1 | - | 4 - 5 | 1030 | 142 | ||
Set Membership: |
|