Search
Search

ARK3| Gene Symbol: ARK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198043.1 | NP_001184972.1 | 15 - 16 | 2534 | 74 | |
| NM_101115.3 | NP_563908.1 | 15 - 16 | 2565 | 74 | ||
| GenBank mRNA | AB290930.1 | - | 15 - 16 | 2339 | 74 | |
| AF159052.1 | - | 15 - 16 | 2529 | 74 | ||
Set Membership: |
|