Search
Search

NUP1| Gene Symbol: NUP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337888.1 | NP_001327263.1 | 9 - 10 | 2395 | 64 | |
| NM_111901.4 | NP_187676.1 | 8 - 9 | 2309 | 64 | ||
| GenBank mRNA | AY093026.1 | - | 8 - 9 | 2194 | 64 | |
| BT010595.1 | - | 8 - 9 | 2121 | 64 | ||
Set Membership: |
|