Search
Search

ATS| Gene Symbol: ATS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123627.3 | NP_199077.1 | 5 - 6 | 866 | 143 | |
| GenBank mRNA | AY048691.1 | - | 5 - 6 | 717 | 143 | |
| BT026053.1 | - | 5 - 6 | 709 | 143 | ||
Set Membership: |
|