Search
Search

AT5G42740| Gene Symbol: AT5G42740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344462.1 | NP_001332180.1 | 6 - 7 | 928 | 74 | |
| NM_001344463.1 | NP_001332179.1 | 5 - 6 | 532 | 74 | ||
| NM_123638.5 | NP_199088.1 | 7 - 8 | 744 | 74 | ||
| GenBank mRNA | AJ419524.1 | - | 7 - 8 | 605 | 74 | |
Set Membership: |
|