Search
Search

AT5G47540| Gene Symbol: AT5G47540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124127.6 | NP_199565.4 | 8 - 9 | 1235 | 95 | |
| GenBank mRNA | AF428296.1 | - | 8 - 9 | 1106 | 95 | |
| BT033107.1 | - | 8 - 9 | 805 | 95 | ||
Set Membership: |
|