Search
Search

AT5G49900| Gene Symbol: AT5G49900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344863.1 | NP_001331233.1 | 3 - 4 | 469 | 76 | |
| NM_124368.4 | NP_199801.2 | 3 - 4 | 384 | 76 | ||
| GenBank mRNA | BT002482.1 | - | 3 - 4 | 224 | 76 | |
| BT010551.1 | - | 2 - 3 | 173 | 76 | ||
Set Membership: |
|