Search
Search

ARP| Gene Symbol: ARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336913.1 | NP_001323447.1 | 10 - 11 | 1571 | 116 | |
| NM_001336914.1 | NP_001323448.1 | 9 - 10 | 1560 | 116 | ||
| NM_129709.5 | NP_181677.1 | 10 - 11 | 1592 | 116 | ||
Set Membership: |
|