Search
Search

GEA6| Gene Symbol: GEA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129575.3 | NP_181546.1 | 1 - 2 | 321 | 101 | |
| GenBank mRNA | AF360157.1 | - | 1 - 2 | 201 | 101 | |
| AY113885.1 | - | 1 - 2 | 120 | 101 | ||
Set Membership: |
|