Search
Search

USO1| Gene Symbol: USO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174845.2 | NP_777270.2 | 1 - 2 | 211 | 61 | |
| GenBank mRNA | BC108112.1 | - | 1 - 2 | 211 | 61 | |
| U14186.1 | - | 1 - 2 | 221 | 61 | ||
Set Membership: |
|