Search
Search

CDH1| Gene Symbol: CDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002763.1 | NP_001002763.1 | 12 - 13 | 1940 | 81 | |
| GenBank mRNA | AY508164.1 | - | 12 - 13 | 1940 | 81 | |
| BC147914.1 | - | 12 - 13 | 2050 | 81 | ||
Set Membership: |
|