Search
Search

ODC1| Gene Symbol: ODC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174130.2 | NP_776555.1 | 2 - 3 | 289 | 95 | |
| GenBank mRNA | BC146218.1 | - | 3 - 4 | 400 | 95 | |
| M92441.1 | - | 2 - 3 | 289 | 95 | ||
Set Membership: |
|