Search
Search

STAR| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174189.3 | NP_776614.2 | 3 - 4 | 446 | 71 | |
| GenBank mRNA | BC110213.1 | - | 3 - 4 | 447 | 71 | |
| Y17259.1 | - | 446 | 71 | |||
Set Membership: |
|