Search
Search

PPP1R1B| Gene Symbol: PPP1R1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174647.2 | NP_777072.1 | 2 - 3 | 483 | 107 | |
| GenBank mRNA | BC118264.1 | - | 2 - 3 | 218 | 107 | |
| M27444.1 | - | 2 - 3 | 483 | 107 | ||
Set Membership: |
|