Search
Search

ASAP1| Gene Symbol: ASAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175821.3 | NP_787015.2 | 29 - 30 | 3396 | 98 | |
| GenBank mRNA | AF112886.1 | - | 29 - 30 | 3396 | 98 | |
| EE241806.1 | - | 1 - 2 | 419 | 98 | ||
Set Membership: |
|