Search
Search

FDX1| Gene Symbol: FDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181011.2 | NP_851354.1 | 3 - 4 | 604 | 76 | |
| GenBank mRNA | BC109849.1 | - | 3 - 4 | 654 | 76 | |
| BT030601.2 | - | 3 - 4 | 483 | 76 | ||
| D00467.1 | - | 504 | 76 | |||
| M11746.1 | - | 3 - 4 | 604 | 76 | ||
| M16934.1 | - | 3 - 4 | 604 | 76 | ||
| S78831.1 | - | 3 - 4 | 594 | 76 | ||
Set Membership: |
|