Search
Search

COPG1| Gene Symbol: COPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037815.2 | NP_001032904.1 | 19 - 20 | 2042 | 69 | |
| GenBank mRNA | BC111322.1 | - | 19 - 20 | 2043 | 69 | |
| X70019.1 | - | 1052 | 69 | |||
Set Membership: |
|