Search
Search

DNM1L| Gene Symbol: DNM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046494.2 | NP_001039959.1 | 17 - 18 | 1819 | 83 | |
| GenBank mRNA | BC112710.1 | - | 17 - 18 | 1820 | 83 | |
Set Membership: |
|