Search
Search

PAGR1| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075632.1 | NP_001069100.1 | 4 - 5 | 1671 | 71 | |
| GenBank mRNA | BC116156.1 | - | 2 - 3 | 553 | 71 | |
| BC118095.1 | - | 4 - 5 | 1671 | 71 | ||
| BC123802.1 | - | 1 - 2 | 1193 | 71 | ||
Set Membership: |
|