Search
Search

CCAR2| Gene Symbol: CCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075298.3 | NP_001068766.2 | 16 - 17 | 2425 | 104 | |
| GenBank mRNA | BC151643.1 | - | 16 - 17 | 2186 | 104 | |
| BT026216.1 | - | 2 - 3 | 733 | 104 | ||
Set Membership: |
|